siRNA Details
Gene Symbol: Gm14812 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033544.2 | 7 | 1634 | |||
GenBank mRNA | AK135730.1 | - | 7 | 1636 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV357684.1 | - | 1 | 74 | Chr. X - 99405331 - 99405564 [-] on Build GRCm38 |
