siRNA Details
Gene Symbol: Gm12000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136731.1 | - | 1 | 183 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB625500.1 | - | 1 | 180 | Chr. 11 - 11695133 - 11696441 [-] on Build GRCm38 |
