siRNA Details
Gene Symbol: 9330158H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015589.1 | 2 | 341 | |||
GenBank mRNA | AK034128.1 | - | 2 | 342 | ||
AK043142.1 | - | 398 | ||||
AK139997.1 | - | 158 | ||||
AK142440.1 | - | 373 | ||||
AK165789.1 | - | 1246 |
