siRNA Details
Gene Symbol: 9330158H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015589.1 | 2 | 144 | |||
GenBank mRNA | AK034128.1 | - | 2 | 145 | ||
AK043142.1 | - | 201 | ||||
AK142440.1 | - | 176 | ||||
AK165789.1 | - | 1049 |
