siRNA Details
Gene Symbol: Gm12863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142602.1 | - | 1, 2 | 130 | ||
BB506114.2 | - | 1, 2 | 140 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI155191.1 | - | 1, 2 | 27 | Chr. 4 - 118954533 - 118954816 [-] on Build GRCm38 |
