siRNA Details
Gene Symbol: D930028M14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045847.1 | 3 | 743 | |||
NR_045848.1 | 3 | 641 | ||||
GenBank mRNA | AK142847.1 | - | 3 | 744 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D930028M14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045847.1 | 3 | 743 | |||
NR_045848.1 | 3 | 641 | ||||
GenBank mRNA | AK142847.1 | - | 3 | 744 |