siRNA Details
Gene Symbol: Gm15348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033546.1 | 2 | 167 | |||
GenBank mRNA | AK143914.1 | - | 2 | 168 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY732759.1 | - | 2 | 168 | Chr. 8 - 12709910 - 12719127 [-] on Build GRCm38 |
