siRNA Details
Gene Symbol: Il1bos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015474.1 | 1, 2 | 28 | |||
GenBank mRNA | AK035070.1 | - | 1, 2 | 29 | ||
AK076405.1 | - | 1, 2 | 34 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627324.1 | - | 1, 2 | 29 | Chr. 2 - 129369902 - 129374957 [+] on Build GRCm38 |
