siRNA Details
Gene Symbol: 4631405J19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110333.1 | 6 | 1181 | |||
GenBank mRNA | AK028444.1 | - | 2893 | |||
BC100298.1 | - | 6 | 1181 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV229223.2 | - | 3 | 958 | Chr. 2 - 93030596 - 93033408 [+] on Build GRCm38 |
