siRNA Details
Gene Symbol: 4631405J19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110333.1 | 5 | 768 | |||
GenBank mRNA | AK028444.1 | - | 2480 | |||
BC100298.1 | - | 5 | 768 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV229223.2 | - | 2 | 546 | Chr. 2 - 93030596 - 93033408 [+] on Build GRCm38 |
