siRNA Details
Gene Symbol: 4932412D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040522.1 | 2 | 178 | |||
GenBank mRNA | AK131804.1 | - | 2 | 181 | ||
BY085940.1 | - | 2 | 343 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4932412D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040522.1 | 2 | 178 | |||
GenBank mRNA | AK131804.1 | - | 2 | 181 | ||
BY085940.1 | - | 2 | 343 |