siRNA Details
Gene Symbol: 6030408B16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033803.1 | 2 | 979 | |||
GenBank mRNA | AK134453.1 | - | 2 | 960 | ||
AK144445.1 | - | 2 | 979 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6030408B16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033803.1 | 2 | 979 | |||
GenBank mRNA | AK134453.1 | - | 2 | 960 | ||
AK144445.1 | - | 2 | 979 |