siRNA Details
Gene Symbol: Gm11529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033524.1 | 1 | 167 | |||
GenBank mRNA | AK138406.1 | - | 1 | 168 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724101.1 | - | 1 | 168 | Chr. 11 - 96448323 - 96464547 [-] on Build GRCm38 |
