siRNA Details
Gene Symbol: 4933413L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045508.1 | 5 | 583 | |||
GenBank mRNA | AK016808.1 | - | 5 | 584 | ||
BU946227.1 | - | 6 | 728 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716484.1 | - | 5 | 584 | Chr. 13 - 117706767 - 117720011 [-] on Build GRCm38 |
