siRNA Details
Gene Symbol: Gm11468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033467.1 | 1 | 61 | |||
GenBank mRNA | BB870378.1 | - | 1 | 62 | ||
BB872766.1 | - | 1 | 62 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm11468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033467.1 | 1 | 61 | |||
GenBank mRNA | BB870378.1 | - | 1 | 62 | ||
BB872766.1 | - | 1 | 62 |