siRNA Details
Gene Symbol: C030047K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021157.1 | - | 3 | 302 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728322.1 | - | 3 | 299 | Chr. 15 - 18698194 - 18818772 [-] on Build GRCm38 |
