siRNA Details
Gene Symbol: Gm12371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081400.1 | - | 2 | 249 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB648387.1 | - | 2 | 249 | Chr. 4 - 36952966 - 36987589 [+] on Build GRCm38 |
