siRNA Details
Gene Symbol: Ptprtos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040617.1 | 3 | 404 | |||
GenBank mRNA | AK021192.1 | - | 3 | 405 | ||
BB398491.1 | - | 1 | 44 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728759.1 | - | 3 | 402 | Chr. 2 - 162390815 - 162393948 [+] on Build GRCm38 |
