siRNA Details
Gene Symbol: B430010I23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015457.1 | 2 | 409 | |||
GenBank mRNA | AK021016.1 | - | 2 | 140 | ||
AK033372.1 | - | 2 | 409 | |||
AK087094.1 | - | 2 | 140 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB624594.1 | - | 2 | 409 | Chr. 8 - 41017468 - 41022512 [+] on Build GRCm38 |
