siRNA Details
Gene Symbol: LOC105244195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079188.1 | - | 2 | 1309 | ||
AK162487.1 | - | 2 | 1307 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105244195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079188.1 | - | 2 | 1309 | ||
AK162487.1 | - | 2 | 1307 |