siRNA Details
Gene Symbol: Gm15927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051804.1 | - | 1 | 118 | ||
AK142821.1 | - | 1 | 86 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB657467.1 | - | 1 | 118 | Chr. 7 - 3635345 - 3644636 [-] on Build GRCm38 |
