siRNA Details
Gene Symbol: 4930570G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040398.1 | 3 | 318 | |||
NR_040399.1 | 2 | 229 | ||||
GenBank mRNA | AK016265.1 | - | 2 | 229 | ||
AK083124.1 | - | 3 | 310 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930570G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040398.1 | 3 | 318 | |||
NR_040399.1 | 2 | 229 | ||||
GenBank mRNA | AK016265.1 | - | 2 | 229 | ||
AK083124.1 | - | 3 | 310 |