siRNA Details
Gene Symbol: Gm12589 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079017.1 | - | 2 | 189 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB625534.1 | - | 2 | 184 | Chr. 11 - 121803111 - 121810319 [+] on Build GRCm38 |
