siRNA Details
Gene Symbol: 0610005C13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038166.1 | 4 | 525 | |||
GenBank mRNA | AK085337.1 | - | 4 | 524 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 0610005C13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038166.1 | 4 | 525 | |||
GenBank mRNA | AK085337.1 | - | 4 | 524 |