siRNA Details
Gene Symbol: 1700125G02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040651.1 | 2 | 365 | |||
GenBank mRNA | AK007278.1 | - | 2 | 371 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707379.1 | - | 2 | 369 | Chr. 4 - 124890888 - 124894782 [-] on Build GRCm38 |
