siRNA Details
Gene Symbol: E130006D01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045832.1 | 3 | 262 | |||
NR_045833.1 | 3 | 262 | ||||
NR_045834.1 | 3 | 262 | ||||
GenBank mRNA | AK087393.1 | - | 3 | 238 | ||
BC087866.1 | - | 3 | 259 | |||
BX529972.1 | - | 3 | 234 |
