siRNA Details
Gene Symbol: D330050G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040335.1 | 4 | 664 | |||
NR_040336.1 | 3 | 572 | ||||
GenBank mRNA | AK083230.1 | - | 4 | 642 | ||
AK084832.1 | - | 3 | 574 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB659609.1 | - | 3 | 574 | Chr. 2 - 116900152 - 116911700 [+] on Build GRCm38 |
