siRNA Details
Gene Symbol: 2410004I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037963.1 | 3 | 509 | |||
GenBank mRNA | AK010390.1 | - | 3 | 511 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709833.1 | - | 3 | 511 | Chr. 11 - 102958354 - 102964154 [-] on Build GRCm38 |
