siRNA Details
Gene Symbol: Kbtbd8os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005835.1 | - | 2 | 236 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705983.1 | - | 2 | 234 | Chr. 6 - 95114017 - 95117631 [-] on Build GRCm38 |
