siRNA Details
Gene Symbol: 1700047A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019763.1 | - | 3, 4 | 224 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715718.1 | - | 3, 4 | 224 | Chr. 8 - 26082616 - 26089294 [+] on Build GRCm38 |
