siRNA Details
Gene Symbol: 1700020I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015473.1 | 3 | 873 | |||
NR_027832.1 | 3 | 704 | ||||
GenBank mRNA | AK006166.1 | - | 3 | 843 | ||
AK009929.1 | - | 3 | 680 | |||
AK011393.1 | - | 3 | 837 | |||
AK017100.1 | - | 3 | 705 | |||
AK029148.1 | - | 839 | ||||
AK030275.1 | - | 836 | ||||
AK147270.1 | - | 850 | ||||
AK170765.1 | - | 3 | 843 | |||
BC026913.1 | - | 3 | 670 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY710677.1 | - | 3 | 838 | Chr. 2 - 119594332 - 119600200 [+] on Build GRCm38 |
