siRNA Details
Gene Symbol: E330012B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033640.1 | 2 | 145 | |||
GenBank mRNA | AK054296.1 | - | 2 | 144 | ||
AK163309.1 | - | 2 | 144 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E330012B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033640.1 | 2 | 145 | |||
GenBank mRNA | AK054296.1 | - | 2 | 144 | ||
AK163309.1 | - | 2 | 144 |