siRNA Details
Gene Symbol: Gm7854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028417.1 | 8 | 1388 | |||
GenBank mRNA | AK138349.1 | - | 6 | 837 | ||
AV378858.2 | - | 3 | 634 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm7854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028417.1 | 8 | 1388 | |||
GenBank mRNA | AK138349.1 | - | 6 | 837 | ||
AV378858.2 | - | 3 | 634 |