siRNA Details
Gene Symbol: 1700019B21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045442.1 | 3 | 410 | |||
NR_045443.1 | 3 | 257 | ||||
GenBank mRNA | AK006108.1 | - | 3 | 258 | ||
BU936494.1 | - | 3 | 413 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706217.1 | - | 3 | 256 | Chr. X - 62510535 - 62516068 [-] on Build GRCm38 |
