siRNA Details
Gene Symbol: Gm15612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045880.1 | 1 | 223 | |||
GenBank mRNA | AK133241.1 | - | 1 | 224 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB616578.1 | - | 1 | 224 | Chr. 6 - 88842854 - 88843875 [+] on Build GRCm38 |
