siRNA Details
Gene Symbol: Pard3bos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015421.1 | - | 470 | |||
AK076674.1 | - | 3 | 548 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pard3bos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015421.1 | - | 470 | |||
AK076674.1 | - | 3 | 548 |