siRNA Details
Gene Symbol: G630018N14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090209.1 | - | 2 | 210 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728512.1 | - | 2 | 169 | Chr. 13 - 31371354 - 31408882 [-] on Build GRCm38 |
