siRNA Details
Gene Symbol: Gm16731 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK164782.1 | - | 173 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730861.1 | - | 1, 2 | 169 | Chr. 4 - 46535904 - 46539402 [+] on Build GRCm38 |
