siRNA Details
Gene Symbol: AW822252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110489.1 | 6 | 842 | |||
GenBank mRNA | AK082553.1 | - | 6 | 843 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB389450.2 | - | 4 | 378 | Chr. X - 53696922 - 53715404 [-] on Build GRCm38 |
