siRNA Details
Gene Symbol: Gm16159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110510.1 | 2, 3 | 308 | |||
GenBank mRNA | AK133017.1 | - | 2, 3 | 309 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615746.1 | - | 2, 3 | 307 | Chr. 8 - 25553515 - 25556082 [-] on Build GRCm38 |
