siRNA Details
Gene Symbol: Gm16159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110510.1 | 5 | 727 | |||
GenBank mRNA | AK133017.1 | - | 5 | 729 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB015528.2 | - | 3 | 414 | Chr. 8 - 25548671 - 25554575 [-] on Build GRCm38 |
