siRNA Details
Gene Symbol: Kis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003188.1 | 1 | 1979 | |||
GenBank mRNA | AK084356.1 | - | 2 | 1411 | ||
AY940614.1 | - | LIT3387 Kis2 |
1 | 1979 | ||
AY940615.1 | - | LIT3388 Kis2 |
2 | 1665 | ||
AY940616.1 | - | LIT3389 Kis2 |
1 | 1298 | ||
AY940617.1 | - | LIT3390 Kis2 |
2 | 826 | ||
AY940618.1 | - | LIT3391 Kis2 |
3 | 512 |
