siRNA Details
Gene Symbol: 4930519D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045975.1 | 3 | 431 | |||
GenBank mRNA | AK015838.1 | - | 3 | 431 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715382.1 | - | 3 | 431 | Chr. 13 - 30615043 - 30623990 [+] on Build GRCm38 |
