siRNA Details
Gene Symbol: Gm15850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046167.1 | 1 | 34 | |||
NR_046168.1 | 1 | 34 | ||||
NR_046169.1 | 1 | 34 | ||||
NR_046170.1 | 1 | 34 | ||||
GenBank mRNA | AK078387.1 | - | 35 | |||
BC107309.1 | - | 1 | 22 | |||
BC107310.1 | - | 1 | 22 | |||
BC114580.1 | - | 1 | 22 | |||
BC115961.1 | - | 1 | 22 |
