siRNA Details
Gene Symbol: 1700007J10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045476.1 | 5 | 796 | |||
GenBank mRNA | AK005722.1 | - | 5 | 798 | ||
AK019811.1 | - | 3 | 365 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715921.1 | - | 3 | 361 | Chr. 11 - 59726014 - 59732769 [-] on Build GRCm38 |
