siRNA Details
Gene Symbol: E330017L17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045190.2 | 3 | 802 | |||
GenBank mRNA | AK054344.1 | - | 3 | 713 | ||
AK054442.1 | - | 3 | 803 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB556875.2 | - | 3 | 624 | Chr. 4 - 129906404 - 129908997 [+] on Build GRCm38 |
