siRNA Details
Gene Symbol: 1700042O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045178.1 | 4 | 730 | |||
GenBank mRNA | AK006690.1 | - | 4 | 731 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706681.1 | - | 4 | 731 | Chr. 11 - 11868123 - 11885256 [+] on Build GRCm38 |
