siRNA Details
Gene Symbol: Gm12996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078255.1 | - | 2 | 398 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719016.1 | - | 2 | 398 | Chr. 4 - 116824903 - 116830590 [+] on Build GRCm38 |
