siRNA Details
Gene Symbol: Gm16548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037987.1 | 3 | 1169 | |||
GenBank mRNA | AK149537.1 | - | 3 | 942 | ||
BC019478.1 | - | 3 | 1169 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm16548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037987.1 | 3 | 1169 | |||
GenBank mRNA | AK149537.1 | - | 3 | 942 | ||
BC019478.1 | - | 3 | 1169 |