siRNA Details
Gene Symbol: C030005K06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021048.1 | - | 4 | 491 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728108.1 | - | 4 | 488 | Chr. 18 - 50042730 - 50053660 [-] on Build GRCm38 |
